missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Gateway™ BP Clonase™ Enzyme Mix DFS Item

Manufacturer:  Invitrogen™ 11789013

 View more versions of this product

Catalog No. 11789013



Description

Description

  • Proteins catalyze in vitro recombination of PCR products or DNA segments from clones (containing attB sites) and Donor vector (containing attP sites) to generate Entry clones
  • BP Clonase II enzyme format is single mix of both enzyme and buffer to ensure enzyme stability and enable convenient 10μL reaction set up with fewer pipetting steps, which both maximize recombination efficiency and minimize pippetting
  • Original format of BP Clonase enzyme is stand alone enzyme blend and reaction buffer provided in separate tubes

Cloning, Gateway Cloning, Gateway Recombination

Specifications

Specifications

Dry Ice
Reaction Buffer
BP Clonase Enzyme Mix
BP Clonase
20 Reactions
Clonase™, Gateway™
SDS
Documents
Product Certifications
Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.