Please login to your online account to display your discounted pricing

Invitrogen™ ChargeSwitch™-Pro Plasmid Miniprep Kit DFS Item


No harsh or potentially interfering reagents

Manufacturer: invitrogen™  LSCS30050

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. CS30050

  • / Each
 

Description & Specifications

Specifications

Target Plasmid DNA
Sample Size >100 μg (Small-Scale) Plasmid DNA
For Use With (Application) Cloning, In Vitro Transcription, Next-Generation Sequencing, Nucleic Acid Labeling, PCR, Sequencing, Transfection, Transformation
Sample Type Bacterial Culture
Isolation Technology ChargeSwitch™ Chemistry
Quantity 50 preps