Please login to your online account to display your discounted pricing

Invitrogen™ ChargeSwitch™-Pro PCR Clean-Up Kit DFS Item


Complete kit for fast, hassle-free setup

Manufacturer: invitrogen™  LSCS32010

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. CS32010

  • / Each
 

Description & Specifications

Specifications

Target DNA from Enzymatic Reactions (e.g. PCR)
Sample Type PCR Products
For Use With (Application) Cloning, Microarray Analysis, Next-Generation Sequencing, PCR, Sequencing
Isolation Technology ChargeSwitch™ Chemistry
Quantity 10 preps