Please login to your online account to display your discounted pricing

Invitrogen™ ChargeSwitch™ NoSpin Plasmid Micro Kit DFS Item

Optimized for automation

$477.00 - $4,505.00

Specifications

For Use With (Application) Cloning, In Vitro Transcription, Next-Generation Sequencing, Nucleic Acid Labeling, PCR, Sequencing, Transfection, Transformation
Isolation Technology ChargeSwitch™ Chemistry
Sample Type Bacterial Culture
Target Plasmid DNA
View More Specs
Includes:  Kit includes all the necessary reagents for the number of preps listed.
Requires:  Magnetic accessory

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Price Quantity    

CS10201

DFS Item
 
invitrogen™
LSCS10201
Each for $477.00

CS1020110

DFS Item
 
invitrogen™
LSCS1020110
Each for $4,505.00
Description & Specifications

Specifications

For Use With (Application) Cloning, In Vitro Transcription, Next-Generation Sequencing, Nucleic Acid Labeling, PCR, Sequencing, Transfection, Transformation
Isolation Technology ChargeSwitch™ Chemistry
Sample Type Bacterial Culture
Target Plasmid DNA

  • Sample size 0.5-1mL overnight culture; Typical yield up to 5μg
  • Provides centrifuge-free purification, ideal for automation
  • Avoids introduction of enzymatic contaminants for better results
  • ChargeSwitch NoSpin Kits provide centrifuge-free microprep plasmid purification, ideal for automation
  • Novel NoSpin reagent causes bacterial cell flocculation and allows cells to be pelleted without the need for automation bottlenecks such as centrifugation

DNA & RNA Purification & Analysis, DNA Extraction, Miniprep, Plasmid DNA Purification

Shipping Condition: Room Temperature