Please login to your online account to display your discounted pricing

Invitrogen™ ChargeSwitch™ NoSpin Plasmid Micro Kit DFS Item


Optimized for automation

Manufacturer: invitrogen™  LSCS1020110

 View more versions of this product

Magnetic accessory

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. CS1020110

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store as follows: ChargeSwitch™ Precipitation Buffer (N5) at 4°C. Mix the RNase I in ChargeSwitch™ Resuspension Buffer (R4) and store the resulting solution at 4°C. Store all other components at room temperature. All components are guaranteed stable for 6 months if stored properly.

Components:

ChargeSwitch™ MagnaClear™ Beads
ChargeSwitch™ Magnetic Beads
ChargeSwitch™ Resuspension Buffer (R4; 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
ChargeSwitch™ Lysis Buffer (L9)
ChargeSwitch™ Precipitation Buffer (N5)
ChargeSwitch™ Wash Buffer (W11)
ChargeSwitch™ Wash Buffer (W12)
ChargeSwitch™ Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)
RNase A (5 mg/mL in 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
Isolation Technology ChargeSwitch™ Chemistry
Target Plasmid DNA
For Use With (Application) Cloning, In Vitro Transcription, Next-Generation Sequencing, Nucleic Acid Labeling, PCR, Sequencing, Transfection, Transformation
Sample Size >100 μg (Small-Scale) Plasmid DNA
Quantity 960 preps
Sample Type Bacterial Culture