Please login to your online account to display your discounted pricing

Invitrogen™ ChargeSwitch™ gDNA Buccal Cell Kit DFS Item


Kit includes necessary reagents

Manufacturer: invitrogen™  LSCS1102110

 View more versions of this product

Use of a magnetic accessory.

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. CS1102110

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Forensic Testing, PCR
Isolation Technology ChargeSwitch™ Chemistry
Target Genomic DNA
Quantity 960 preps
Sample Type Buccal Samples
Quantity 960 preps