Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ siPORT™NeoFX Transfection Agent, Serum Compatible DFS Item


Lipid-based formulation can be used to efficiently transfect adherent cells with siRNA as they are subcultured—without increased cytotoxicity

Manufacturer: invitrogen™  LSAM4510M

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. AM4510M

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Synthetic siRNA
Quantity 400μL
Includes With manual
Quantity 400μL
Storage Requirements Store at 4°C
Storage Conditions Store at 4°C