missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ RNA Century™-Plus Marker Templates DFS Item

Contain mixtures of linearized plasmids ready for use as templates during in vitro transcription reactions for synthesis of labeled RNA molecular size standards

Manufacturer:  Invitrogen™ LSAM7782

Catalog No. AM7782



Description

Description

When transcribed with T7 RNA polymerase, the templates produce 7 RNA transcripts with lengths of 100, 200, 300, 400, 500, 750, and 1000 bases. They can be labeled with radioisotopes, biotin, or other nonisotopic modified nucleotides. They are ideal as size standards for ribonuclease protection assays and other applications where a low molecular weight RNA ladder is useful. One tube contain 5 μg (0.5mg/mL).

Acrylamide Gel Electrophoresis, DNA & RNA Purification & Analysis, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Conditions: Room Temperature

Specifications

Specifications

Nucleic Acid Gel Electrophoresis & Blotting
RNA Marker
Store at –20°C.
No
0.1 to 1 kb
Unconjugated
0.5 mL
Denaturing Polyacrylamide Gels
Room Temperature
Ambion™, Century™
SDS
Documents

For Research Use Only. Not for use in diagnostic procedures.