missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ RNA Century™ Marker Templates DFS Item

Contain mixtures of linearized plasmids ready for use as templates as part of in vitro transcription reactions for synthesis of labeled RNA molecular size standards

Manufacturer:  Invitrogen™ LSAM7780

Catalog No. AM7780



Description

Description

When transcribed with T7 RNA polymerase, the templates produce five RNA transcripts with the lengths of 100, 200, 300, 400, and 500 bases. They can be labeled with radioisotopes, biotin, or other nonisotopic modified nucleotides. They are ideal as size standards for ribonuclease protection assays and other applications where a low molecular weight RNA ladder is useful.

Acrylamide Gel Electrophoresis, DNA & RNA Purification & Analysis, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Conditions: Room Temperature

Specifications

Specifications

Unconjugated
Denaturing Polyacrylamide Gels
Room Temperature
Ambion™, Century™
Store at –20°C.
No
100 to 500 bp
SDS
Documents

For Research Use Only. Not for use in diagnostic procedures.