missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ pUC19 DNA (Sau3A I digested)
Ambion pUC 19 DNA is digested to completion with Sau3A I
Manufacturer: Invitrogen™ LSAM7760
Description
Yields fragments of 955, 585, 341, 258, 141, 105, 78, 75, 46, 36, 18, 17, 12, 11, and 8 bp. Markers are visualized by staining with ethidium bromide. Alternatively, they may be radioactively end-labeled with Exo- Klenow or T4 Polynucleotide Kinase. One tube containing 50μg is provided.
Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting
Order Info
Shipping Conditions: Room Temperature
Specifications
0.5 mg/ml, 0.5 mg⁄ml, 0.5 mg/ml | |
Denaturing Polyacrylamide Gels, Agarose Gels | |
50 µg | |
Room Temperature | |
Ambion™ |
Store at –20°C. | |
0.5 mg⁄ml | |
No | |
8 to 955 bp |
For Research Use Only. Not for use in diagnostic procedures.