AccuPrime™ GC-Rich DNA Polymerase DFS Item
High-specificity, high-yield PCR amplification of GC-rich templates
Manufacturer:  Life Technologies LS12337024
 View more versions of this product
Catalog No. 12337024
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
AccuPrime GC-Rich DNA Polymerase is supplied with two 5X reaction buffers and a separate vial of 50mM MgSO4 . Both buffers contain dNTPs, MgSO4 , and AccuPrime accessory proteins. Reaction buffer A is for genomic targets. Reaction buffer B is for use with cDNA, plasmid, or lambda targets.
Gel Compatibility: High
Store at -20°C.
1000 reactions
Fidelity (vs. Taq): 2X