missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Platinum™ SuperFi™ Green PCR Master Mix DFS Item

$7.00 - $1,562.00

Specifications

Storage Requirements Store at -20°C.
Includes 2X Platinum SuperFi Green PCR Master Mx, 5X SuperFi GC Enhancer, nuclease-free water.
For Use With (Application)
  • High-fidelity PCR
  • Cloning and sub-cloning
  • Site-directed mutagenesis
  • Amplification of GC-rich templates 
  • Template generation for sequencing
  • High-throughput PCR
  • Amplification of samples with suboptimal purity
  • Long PCR (up to 20 kb)
  • Fast PCR
Format 2X master mix
Overhang Blunt
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
12359002 Life Technologies
LS12359002
20 rxns Each for $7.00
12359010 Invitrogen™
LS12359010
100 rxns Each for $341.00
12359050 Invitrogen™
LS12359050
500 rxns Each for $1,562.00
Specifications

Description

Invitrogen™ Platinum™ SuperFi™ DNA Polymerase is a proofreading DNA polymerase which combines superior fidelity with trusted PlatinumTM hot-start technology for the highest success in PCR. Featuring >100x Taq fidelity, Platinum SuperFi DNA Polymerase is ideally suited for cloning, mutagenesis and other applications benefiting from supreme sequence accuracy.

  • Error-free PCR products due to exceptional >100X Taq fidelity
  • High specificity and increased yields with Platinum hot-start technology
  • Robust amplification of difficult-to-amplify targets including those of sub-optimal purity or with ˃65% GC content 
  • Convenient workflow with room temperature reaction setup and direct gel loading with green master mix format
Description & Specifications

Specifications

Store at -20°C.
  • High-fidelity PCR
  • Cloning and sub-cloning
  • Site-directed mutagenesis
  • Amplification of GC-rich templates 
  • Template generation for sequencing
  • High-throughput PCR
  • Amplification of samples with suboptimal purity
  • Long PCR (up to 20 kb)
  • Fast PCR
Blunt
2X Platinum SuperFi Green PCR Master Mx, 5X SuperFi GC Enhancer, nuclease-free water.
2X master mix