Please login to your online account to display your discounted pricing

Platinum™ SuperFi™ Green PCR Master Mix DFS Item


Manufacturer: life technologies  LS12359002

 View more versions of this product

Catalog No. 12359002

  • / Each
 

Description & Specifications

Specifications

Quantity 20 rxns
For Use With (Application)
  • High-fidelity PCR
  • Cloning and sub-cloning
  • Site-directed mutagenesis
  • Amplification of GC-rich templates 
  • Template generation for sequencing
  • High-throughput PCR
  • Amplification of samples with suboptimal purity
  • Long PCR (up to 20 kb)
  • Fast PCR
Includes 2X Platinum SuperFi Green PCR Master Mx, 5X SuperFi GC Enhancer, nuclease-free water.
Overhang Blunt
Format 2X master mix
Storage Requirements Store at -20C.

Invitrogen™ Platinum™ SuperFi™ DNA Polymerase is a proofreading DNA polymerase which combines superior fidelity with trusted PlatinumTM hot-start technology for the highest success in PCR. Featuring >100x Taq fidelity, Platinum SuperFi DNA Polymerase is ideally suited for cloning, mutagenesis and other applications benefiting from supreme sequence accuracy.