Invitrogen™ 123 bp DNA Ladder DFS Item
Suitable for sizing double-stranded DNA from 123 to 3075bp
$85.31 - $85.31
Specifications
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
15613011 Life Technologies
LS15613011
100μL Each for $85.31
Specifications
Description
Prepared from a plasmid containing repeats of a 123-bp DNA fragment (1), the ladder consists of 34 fragments ranging in length from 123 to 4,182bp. Up to 25 separate bands may be clearly visualized on 0.7% to 1.5% agarose gels after ethidium bromide staining. The ladder is also useful on 4% to 8% polyacrylamide gels. This ladder can be stained with ethidium bromide or radiolabeled using T4 polynucleotide kinase, T4 DNA polymerase, DNA polymerase I, or the large fragment of DNA polymerase I (Klenow fragment). Compatible with agarose gels.
Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Approved for shipment on Wet or Dry Ice
Description & Specifications
Specifications
0.0