Investigating Water Pollutants and Water Analysis
Learn how to analyze local water samples.
Supplier: Fisher Science Education AZ201503
Includes: Teacher and student guides
Description
- Use these safe chemical tests and test strips to learn the basics of water pollution
- Test for a variety of common pollutants such as total and free chlorine, total hardness, total alkalinity and pH
- Quantify the concentration of pollutants to determine the potential danger
Order Info
Class size: 40 students.
Specifications
Learn how to analyze local water samples | |
Teacher and student guides | |
40 Students |