Please login to your online account to display your discounted pricing

Investigating Spontaneous Generation


Recreate Pasteur's classic experiment.

Manufacturer: fisher scientific  AZ-20-1315

 View more versions of this product

Catalog No. S32621

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Students can recreate pasteur's findings
Product Type Investigating Spontaneous Generation
Kit Size Refill Pack
Class Size Refill Pack