Please login to your online account to display your discounted pricing

Investigating Spontaneous Generation

Recreate Pasteur's classic experiment.

$32.40 - $32.40

Specifications

For Use With (Application) Students can recreate pasteur's findings
Product Type Investigating Spontaneous Generation
View More Specs
Products
Catalog Number Mfr. No. Kit Size Price Quantity    

S32621

 
fisher scientific
AZ-20-1315
Refill Pack Each for $32.40
Description & Specifications

Specifications

For Use With (Application) Students can recreate pasteur's findings
Product Type Investigating Spontaneous Generation

Can life arise from nonlife? Use this kit to recreate Pasteur's experiment and give students firsthand knowledge into the fundamentals of evolution. Compares the growth of organisms exposed to particles in the atmosphere with those exposed to air but not particulate matter.

Class size: 40 students