Please login to your online account to display your discounted pricing

Investigating Sound Waves


Use tuning forks to explore the concepts of sound.

Manufacturer: fisher science education  AZ-20-1643

 View more versions of this product

Catalog No. S65141

  • / Each
 

Description & Specifications

Specifications

Includes Tuning forks
Kit Size 40 students
For Use With (Application) Used to investigate sound waves
Class Size 40 students
Product Type Investigating Sound Waves Kit