Please login to your online account to display your discounted pricing

Investigating Sound Waves

Use tuning forks to explore the concepts of sound.

$156.70 - $156.70

Specifications

Class Size 40 students
For Use With (Application) Used to investigate sound waves
Includes Tuning forks
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    

S65141

 
fisher science education
AZ-20-1643
Investigating Sound Waves Kit Each for $156.70
Description & Specifications

Specifications

Class Size 40 students
For Use With (Application) Used to investigate sound waves
Includes Tuning forks

  • Interaction of waves
  • Vibration of sound in a medium
  • Sympathetic vibration
  • Resonance
  • Interference
  • Doppler effect

Curriculum pack includes kit and corresponding CD-ROM: Waves and Sound, single user (S65007).

Class size: 40 students.