missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Investigating Physical Science Poster Set

Graphic guides to key physical science concepts.

$151.20 - $151.20

Specifications

Classification Science
For Use With (Application) Detailed illustrations depicting key physical science topics
Height (English) 35 in.
Height (Metric) 88.9cm
Includes Posters on investigating force and motion, heat, sound, light, magnetism
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Material Price Quantity  
Catalog Number Mfr. No. Material Price Quantity  
S01456 Fisher Science Education
35-1136
Laminated Each for $151.20
Specifications

Description

Eye-catching reference charts feature detailed illustrations depicting key physical science topics.

  • Sized at a large 23 x 35 in.
  • Posters are offered in either high gloss paper or are laminated for years of classroom use
  • Set of 6, covering topics which include Force and Motion, Heat, Sound, Light, and Magnetism
Description & Specifications

Specifications

Science
35 in.
Posters on investigating force and motion, heat, sound, light, magnetism
58.42cm
Detailed illustrations depicting key physical science topics
88.9cm
23 in.