Investigating Animal Behavior
Study the response of organisms to environmental conditions.
Manufacturer:  Fisher Scientific AZ992316
 View more versions of this product
Catalog No. S85302
Includes: Eight choice chambers and prepared solutions to test various environmental conditions; additional choice chambers available
Description
Description
In this lab investigation, students use specially molded choice chambers to study the response of organisms to environmental conditions.
  • Determine the environmental preference of locally available animals, such as sow bugs, ants, earthworms and other small invertebrates
  • Test students' predictions regarding habitat preference, as well as the innate and learned behaviors of animals
Specifications
Specifications
Students use specially molded choice chambers to study the response of organisms to environmental conditions
Eight choice chambers and prepared solutions to test various environmental conditions, additional choice chambers available
Choice Chamber
7.5 x 7.5 x 0.87 in. (19.1 x 19.05 x 2.2cm)