Please login to your online account to display your discounted pricing

Introduction to Chromatography Lab


Separate plant pigments and dye samples to learn about chromatography in clinical, forensic and analytical laboratories.

Manufacturer: fisher science education  201685

 View more versions of this product

Catalog No. S67397

  • / Each
 

Description & Specifications

Specifications

Class Size 40 students
For Use With (Application) Separates plant pigments and dye samples, employed to identify organic and inorganic compounds
Product Type Lab Investigation Kit