Purus™ International Latex Cleanroom Paper, 22 lb.: PROMO
Write, print and photocopy in cleanroom environments where the prevention of chemical exportation and particle contamination is critical.
Manufacturer:  Purus™ LCIB2021CPM
 View more versions of this product
Catalog No. 19-125-911PM
Description
Description
  • Features high tensile strength and excellent heat resistance for enhanced copier and printing efficiency
  • Polyethylene binding technology results in very low levels of particles, nonvolatile residue (NVR) and ion extractables
  • Unique precision cutting technology guarantees consistent sizing and cleanliness
  • Dimensions (W x L) 21.6cm (8.5 in.) x 27.9cm (11 in.)
  • Weight 9kg (22 lb.)
  • Paper is web-cleaned on both sides prior to sheeting
Specifications
Specifications
Blue
For writing, printing, photocopying in cleanroom environments
279mm
279mm
9.97kg
216mm
11 x 8.5 x 11 in. (279 x 216 x 279mm)
11 in.
11 in.
22 lb.
8.5 in.
Cleanroom Paper