International Enviroguard™ MicroGuard CE™ Clean Processed Microporous Boot Covers
Disposable boot covers are made from high MVTR, innovative fabric. Ideal for ISO 5 or higher non-hazardous work environments where reliable liquid and particulate protection is needed.
Supplier:  International Enviroguard™ CE8104CIL
 View more versions of this product
Catalog No. 19-472-558
Description
Description
These microporous boot covers are clean processed to offer the perfect balance of safety and comfort. The fabric keeps dust and liquids out while allowing air to flow freely, so your feet stay cool throughout a hard day's work. These disposable boot covers are ideal for non-hazardous work environments, such as controlled environments or cleanrooms, where you need reliable liquid and particulate protection. Boot covers are designed with tunnelized elastic to contain particle shed. Non-skid soles provide added traction to help prevent accidental slips and trips. Each boot cover is individually packaged to help prevent cross-contamination.
  • Non-skid soles help improve traction to prevent accidental slips
  • Elastic closures for snug, secure fit that keeps boot covers in place
  • Available in Bulk or Individually packaged to prevent cross-contamination
Specifications
Specifications
Large
White
Non-skid
28 mil
200/Cs.
17 in.
Controlled Environments Boot Covers
Microporous Fabric
Elastic Top and Ankles
Serged
ANSI ISEA 101-1996 R2020, ASTM T903, ASTM F1670-07
Individually Packaged
43.18 cm