Please login to your online account to display your discounted pricing

Internal/Surrogate Standard Mixture (For EPA 524.2)

$113.71 - $113.71

Chemical Identifiers

Chemical Name or Material Internal/Surrogate Standard Mixture
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

US-STM-320N-1

 
ultra scientific
STM320N1
Ampule 1x1mL Each for $113.71
Description & Specifications

Specifications

Quantity 1x1mL
Concentration or Composition (by Analyte or Components) Each component at 2000μg/mL in Methanol:
4-Bromofluorobenzene
1, 2-Dichlorobenzene-d4
Fluorobenzene
Methods For EPA Method 524.2
Packaging Ampule
Chemical Name or Material Internal/Surrogate Standard Mixture

Page 301C, Designates A2LA-Certified product