missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Internal Standard Mixture, Ultra Scientific

$110.77 - $110.77

Chemical Identifiers

Physical Form Liquid
Chemical Name or Material Internal Standard Mixture, 3 analytes at 2000μg/mL in water
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
US-STM-250N-1 Ultra Scientific
STM250N1
Ampule 1x1mL Each for $110.77
Specifications

Description

Page 300C, Designates A2LA-Certified product

Description & Specifications

Chemical Identifiers

Liquid
Internal Standard Mixture, 3 analytes at 2000μg/mL in water

Specifications

1x1mL
Liquid
Ampule
Each component at 2000μg/mL in Methanol:
1, 2-Dichlorobenzene-d4
Fluorobenzene
1
Internal Standard Mixture, 3 analytes at 2000μg/mL in water