Please login to your online account to display your discounted pricing

Internal Standard Mixture (For EPA 8260)

$145.77 - $145.77

Chemical Identifiers

Chemical Name or Material Internal Standard Mixture
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

US-STM-341N-1

 
ultra scientific
STM341N1
Ampule 1x1mL Each for $145.77
Description & Specifications

Specifications

Quantity 1x1mL
Concentration or Composition (by Analyte or Components) Each component at 2000μg/mL in Methanol:
Chlorobenzene-d5
1, 4-Difluorobenzene
1, 4-Dichlorobenzene-d4
Pentafluorobenzene
Methods For EPA Method 8260
Chemical Name or Material Internal Standard Mixture
Packaging Ampule

Page 311C, Designates A2LA-Certified product