Please login to your online account to display your discounted pricing

Internal Standard, SPEX CertiPrep™


ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Quantity 2mL
Packaging 2mL amber prescored glass ampule

Manufacturer: spex certiprep llc  CLPSI2

 View more versions of this product

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Catalog No. CLPS-I2

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 2mL
DOT Information Non-Hazardous
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Methods For US EPA Methods 625, 8270C, CLP
Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Solvent or Matrix MeCl
Name Note EPA Method 625, SW-846 Method 8270C, CLP Methods
Concentration or Composition (by Analyte or Components) 4000μg/mL
CAS Various
Chemical Name or Material Internal Standard
Recommended Storage Ambient

Specifications

Chemical Name or Material Internal Standard
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,4-Dichlorobenzene-d43855-82-12000μg/mL
Acenaphthene-d1015067-26-22000μg/mL
Chrysene-d121719-03-52000μg/mL
Naphthalene-d81146-65-22000μg/mL
Perylene-d121520-96-32000μg/mL
Phenanthrene-d101517-22-22000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Ambient