missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Internal Standard, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$264.20 - $289.51

Chemical Identifiers

For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Chemical Name or Material Internal Standard
View More Specs

Disclaimers:

Product intended for laboratory use only


Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CLPS-I2 Spex Certiprep Llc
CLPSI2
2mL amber prescored glass ampule 2mL Each for $289.51
CLPS-I Spex Certiprep Llc
CLPSI
2mL amber prescored glass ampule 1mL Each for $264.20
Specifications

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,4-Dichlorobenzene-d43855-82-14000μg/mL
Acenaphthene-d1015067-26-24000μg/mL
Chrysene-d121719-03-54000μg/mL
Naphthalene-d81146-65-24000μg/mL
Perylene-d121520-96-34000μg/mL
Phenanthrene-d101517-22-24000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Internal Standard

Specifications

2mL
For US EPA Methods 625, 8270C, CLP
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
4000μg/mL
Internal Standard, EPA Method 625, SW-846 Method 8270C, CLP Methods
GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
2mL amber prescored glass ampule
MeCl
Various

Safety and Handling

DOTInformation : Non-Hazardous

Recommended Storage : Ambient