Please login to your online account to display your discounted pricing
Intermediate Spectrometer
Robust enough for student use, yet capable of advanced work.
Manufacturer: fisher science education  FSCI030
 View more versions of this product
Catalog No. S96204
  • / Each
Description & Specifications
Specifications
Product Type Intermediate spectrometer
Includes 15X Ramsden eyepiece, Glass cross line graticule, scale, telescope, collimator, prism table
For Use With (Application) For student use capable of advanced work
Scale Type Fixed