Please login to your online account to display your discounted pricing

Intermediate Spectrometer

Robust enough for student use, yet capable of advanced work.

$58.20 - $1,081.35

Specifications

For Use With (Application) For student use capable of advanced work
Scale Type Fixed
View More Specs
Includes:  15X Ramsden eyepiece, glass cross-line graticule
Products
Catalog Number Mfr. No. Includes Product Type Price Quantity    

S96203

 
fisher scientific
PH0619
150mm Table and telescope independent, collimator, 178mm focus achromatic adjustment, 32mm clear aperture, prism table mountable on spectrometer, with 15X Ramsden eyepiece, Glass cross-line graticule Standard spectrometer Each for $1,081.35

S96204

 
fisher science education
FSCI030
15X Ramsden eyepiece, Glass cross line graticule, scale, telescope, collimator, prism table Intermediate spectrometer Each for $58.20
Description & Specifications

Specifications

For Use With (Application) For student use capable of advanced work
Scale Type Fixed

Scale

  • Fixed 150mm diameter
  • Table and telescope independent of each other
  • Both movements can be read to 30 sec. of arc using double verniers
  • Both rotations have fine-adjustment screws
  • Release of clamping screw enables coarse adjustment to be made by hand

Telescope
  • Mounted on movable pillar fitted with 178mm focus achromatic objective
  • 32mm clear aperture
  • Fitted with a 15X Ramsden eyepiece and glass cross-line graticule

Collimator
  • Mounted on movable pillar
  • Fitted with 178mm focus achromatic objective, 32mm clear aperture

Prism Table
  • Marked with lines to assist placing the prism with respect to the table leveling screws
  • Interchangeable clamping units
  • Diffraction grating holder

Supplied in a Styrofoam™ case housed within a wooden case.