Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Interferon-α (II)/IFN-α (II), Mouse Monoclonal Antibody

Recommended for blocking studies, Thermo Scientific™ Interferon-α (II)/IFN-α (II), Mouse Monoclonal Antibody provides accurate, reproducible results.

$243.39 - $758.24

Specifications

Antigen Interferon-alpha (II)/IFN-alpha (II)
Clone N27
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS574P

 
thermo scientific™ lab vision™
MS574P
200μg/mL 1mL Each for $675.59

MS574P0

 
thermo scientific™ lab vision™
MS574P0
200μg/mL 100µL Each for $243.39

MS574P1

 
thermo scientific™ lab vision™
MS574P1
200μg/mL 500µL Each for $475.27

MS574P1ABX

 
thermo scientific™ lab vision™
MS574P1ABX
1mg/mL 100µL Each for $603.16

MS574PABX

 
thermo scientific™ lab vision™
MS574PABX
1mg/mL 200µL Each for $758.24
Description & Specifications

Specifications

Antigen Interferon-alpha (II)/IFN-alpha (II)
Clone N27
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Cytokines
Type Primary
Immunogen Purified recombinant IFN-alpha 2c
Isotype IgG2a kappa

Recommended for blocking studies, Thermo Scientific™ Interferon-α (II)/IFN-α (II), Mouse Monoclonal Antibody provides accurate, reproducible results. 

The IFN-α family consists of 24 or more genes or pseudo-genes. IFN-αII) is one of the two distinct families (I and II) of human IFN-α. The α interferons are mainly produced by lymphocytes, monocytes, macrophages, and cell lines such as Namalwa and KG1 following induction by viruses, nucleic acids, and glucocorticoid hormones. They are involved in virus resistance on target cells, inhibition of cell proliferation, induction of cytokines and regulation of expression of MHC class I antigens.

Host Species: Mouse

Clone: N27

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: aa 43-53

Immunogen: Purified recombinant IFN-α 2c

Molecular Weight: 16-27kDa

Positive Control: Stimulated human peripheral blood mononuclear cells

Recommended for:

  • Blocking study

Note:

USA: RUO; Int'l: RUO