Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Interferon-α (II)/IFN-α (II), Mouse Monoclonal Antibody


Recommended for blocking studies, Thermo Scientific™ Interferon-α (II)/IFN-α (II), Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer: thermo scientific™ lab vision™  MS574P1ABX

 View more versions of this product

Catalog No. MS574P1ABX

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Antigen Interferon-alpha (II)/IFN-alpha (II)
Format Concentrated
Isotype IgG2a kappa
Cross Reactivity Human
Host Species Mouse
Immunogen Purified recombinant IFN-alpha 2c
Quantity 100µL
Clone N27
Conjugate Unlabeled
Research Discipline Cytokines
Type Primary
Classification Monoclonal
Concentration 1mg/mL

Recommended for blocking studies, Thermo Scientific™ Interferon-α (II)/IFN-α (II), Mouse Monoclonal Antibody provides accurate, reproducible results. 

The IFN-α family consists of 24 or more genes or pseudo-genes. IFN-αII) is one of the two distinct families (I and II) of human IFN-α. The α interferons are mainly produced by lymphocytes, monocytes, macrophages, and cell lines such as Namalwa and KG1 following induction by viruses, nucleic acids, and glucocorticoid hormones. They are involved in virus resistance on target cells, inhibition of cell proliferation, induction of cytokines and regulation of expression of MHC class I antigens.

Host Species: Mouse

Clone: N27

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: aa 43-53

Immunogen: Purified recombinant IFN-α 2c

Molecular Weight: 16-27kDa

Positive Control: Stimulated human peripheral blood mononuclear cells

Recommended for:

  • Blocking study

Note:

USA: RUO; Int'l: RUO