Please login to your online account to display your discounted pricing

Integra™ Miltex™ Surgical Instrument Cleaner


Neutral pH (7) detergent

Manufacturer: integra™ miltex™  MLT3725

 View more versions of this product

Catalog No. 12-460-424

  • / Case
 

Description & Specifications

Specifications

Format Liquid
For Use With (Application) To remove organic material from instruments
pH Range 7
Container Type Pump bottle
Size 1 gal.