Please login to your online account to display your discounted pricing

Integra™ Miltex™ Sterile Surgical Blades


Single-use blades for convenience

Manufacturer: integra™ miltex™  MLT4322

 View more versions of this product

Catalog No. 12-460-449

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Size 22
Length (English) 2.24 in.
For Use With (Equipment) For No. 3 or No. 4 scalpel handles
Shelf Life 5 Year(s)
Type Surgical Blade
Disposable Yes
For Use With (Application) For superior cutting and clean incisions
Length (Metric) 5.7cm
Sterile Yes
Material Stainless Steel