Please login to your online account to display your discounted pricing

Integra™ Miltex™ Sterile Surgical Blades


Single-use blades for convenience

Manufacturer: integra™ miltex™  MLT4315

 View more versions of this product

Catalog No. 12-460-448

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Size 15
Type Surgical Blade
Shelf Life 5 Year(s)
Disposable Yes
For Use With (Equipment) For No. 3 or No. 4 scalpel handles
For Use With (Application) For superior cutting and clean incisions
Sterile Yes
Material Stainless Steel
Length (Metric) 3.7cm
Length (English) 1.45 in.
Accessories

5 Year(s)