Please login to your online account to display your discounted pricing

Integra™ Miltex™ Sterile Surgical Blades


Single-use blades for convenience

Manufacturer: integra™ miltex™  MLT4111

 View more versions of this product

Catalog No. 12-460-441

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Size 11
Length (English) 1.57 in.
Length (Metric) 4cm
Type Surgical Blade
Material Carbon Steel
Disposable Yes
For Use With (Application) For superior cutting and clean incisions
For Use With (Equipment) For No. 3 or No. 4 scalpel handles
Shelf Life 5 Year(s)
Sterile Yes