Please login to your online account to display your discounted pricing

Integra™ Miltex™ Sterile Surgical Blades


Single-use blades for convenience

Manufacturer: integra™ miltex™  MLT4110

 View more versions of this product

Catalog No. 12-460-440

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Size 10
For Use With (Equipment) For No. 3 or No. 4 scalpel handles
Disposable Yes
Length (English) 1.57 in.
Type Surgical Blade
For Use With (Application) For superior cutting and clean incisions
Shelf Life 5 Year(s)
Sterile Yes
Material Carbon Steel
Length (Metric) 4cm
Accessories

5 Year(s)