Please login to your online account to display your discounted pricing

Integra™ Miltex™ Sterile Standard Scalpels


Stainless steel, razor-sharp cutting edge for superior cutting and clean incisions

Manufacturer: integra™ miltex™  MLT4411

 View more versions of this product

Catalog No. 12-460-452

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Type Standard Scalpel
Material Stainless Steel
Sterile Yes
Disposable Yes
For Use With (Application) Superior cutting and clean incisions
Shelf Life 5 Year(s)
Size 11