Please login to your online account to display your discounted pricing

Integra™ Miltex™ Standard Biopsy Punches


Stainless-steel razor sharp cutting edge

Manufacturer: integra™ miltex™  MLT3337

 View more versions of this product

Catalog No. 12-460-413

  • / Pack of 50 Each
 

Description & Specifications

Specifications

Diameter (English) 0.31 in.
Diameter (Metric) 0.8cm
For Use With (Application) Offer the convenience of single-use disposability without sacrificing quality, feel or performance
Disposable Yes
Handle Type Ribbed
Material Stainless Steel
Seam Type Seamless
Shelf Life 5 Year(s)
Sterile Yes
Product Type Biopsy Punch