Please login to your online account to display your discounted pricing

Integra™ Miltex™ Scalpel Handles


Manufacturer: integra™ miltex™  MLT420

 View more versions of this product

Catalog No. 12-460-146

  • / Each
 

Description & Specifications

Specifications

Material Plastic
Length (English) Overall 5.25 in.
Length (Metric) Overall 13.3cm
Autoclavable Yes
Color Red
For Use With (Equipment) 20 to 25 Blades
Reusable Yes
Size No. 6
Product Type Scalpel Handle