Integra™ Miltex™ Pencil-Type Micro Scissors
Delicate tips
$2,216.49 - $2,216.49
Specifications
Blade Style Straight
For Use With (Application) Surgical, Precision
Length (English) 7.25 in.
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Length (Metric) Price Quantity  
Catalog Number Mfr. No. Length (Metric) Price Quantity  
12-460-227 Integra™ Miltex™
MLT17115
18.41cm Each for $2,216.49
12-460-233 Integra™ Miltex™
MLT17215
18.5cm Each for $2,216.49
Specifications
Description
  • High-quality surgical-grade stainless steel construction provides optimal performance
  • Fully tempered precision screw locks maintain perfect alignment
  • Ergonomic design combines weight, balance and feel for optimal comfort and performance
  • Well rounded and polished ring handles provide exceptional comfort and reduced hand fatigue
  • 4mm blades
  • Length: 7.25 in. (18.4cm)
Description & Specifications
Specifications
Straight
Surgical, Precision
7.25 in.