Please login to your online account to display your discounted pricing

Integra™ Miltex™ Operating Room Scissors


For general utility applications

Manufacturer: integra™ miltex™  MLT522

 View more versions of this product

Catalog No. 12-460-153

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Surgical, Precision
Material Stainless Steel
Grade Premium
Blade Style Straight
Length (Metric) 11.43cm
Length (English) 4.5 in.
Type Surgical, precision scissors
Tip Style Blunt/Blunt