Please login to your online account to display your discounted pricing

Integra™ Miltex™ Operating Room Scissors


For general utility applications

Manufacturer: integra™ miltex™  MLT519

 View more versions of this product

Catalog No. 12-460-152

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Surgical, Precision
Type Surgical, precision scissors
Blade Style Straight
Material Stainless Steel
Tip Style Sharp/Blunt
Grade Premium
Length (English) 7.5 in.
Length (Metric) 19cm