Please login to your online account to display your discounted pricing

Integra™ Miltex™ Operating Room Scissors


For general utility applications

Manufacturer: integra™ miltex™  MLT514

 View more versions of this product

Catalog No. 12-460-151

  • / Each
 

Description & Specifications

Specifications

Type Surgical, precision scissors
Grade Premium
Length (English) 5 in.
Material Stainless Steel
For Use With (Application) Surgical, Precision
Tip Style Sharp/Blunt
Blade Style Straight
Length (Metric) 12.7cm