Please login to your online account to display your discounted pricing

Integra™ Miltex™ Operating Room Scissors


For general utility applications

Manufacturer: integra™ miltex™  MLT58

 View more versions of this product

Catalog No. 12-460-149

  • / Each
 

Description & Specifications

Specifications

Blade Style Straight
Material Stainless Steel
Tip Style Sharp/Sharp
Grade Premium
Length (English) 6.5 in.
Length (Metric) 16.51cm
Type Surgical, precision scissors
For Use With (Application) Surgical, Precision