missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Integra™ Miltex™ Mid Grade Iris Tissue Forceps

1 x 2 teeth

$40.71 - $42.17

Specifications

Length (English) 4 in.
Length (Metric) 10.2cm
Material Stainless Steel
Product Type Iris Tissue Forceps
Quantity (No. per Case) Each
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Tip Style Price Quantity  
Catalog Number Mfr. No. Tip Style Price Quantity  
12-460-769 Integra™ Miltex™
MLTV918788
Half Curved Each for $42.17
12-460-770 Integra™ Miltex™
MLTV918790
Full Curved Each for $40.71
Specifications

Description

  • Pattern length 4 in. (10.2cm)
  • Stainless steel construction

Description & Specifications

Specifications

4 in.
Stainless Steel
Each
1 x 2
10.2cm
Iris Tissue Forceps
Mid