Integra™ Miltex™ Microsurgical Kit
Kit with a selection of commonly used surgical tools
Supplier: Integra™ Miltex™ 6890
Requires :
Kit includes one each:
- DERF needle holder with grated jaws, 4000 teeth per square in
- 4 in. long (10.2cm) Iris scissors
- 4 in. long (10.2cm) Swiss jeweler style forceps, nonmagnetic stainless steel, style 5, micro-fine
- 3.5 in. long (8.9cm) McPherson micro iris suturing forceps, straight, smooth 0.3 x 5mm platform, no teeth
- 3.25 in. long (8.3cm) McPherson-Vannas micro Iris scissors, straight with sharp points
Description
Available with or without stainless-steel storage cassette and mat.
†Supplied only with Cat. No. 12-460-112.
Specifications
Microsurgical Lab Animal Kit | |
Lab animal | |
DERF needle holder, grated jaw, 4000 teeth, 4 in. L Iris scissor, 4 in. L swiss jeweler style forceps, 3.5 in. L McPherson micro iris suturing forceps, 3.25 in. L McPherson-vannas micro Iris scissors, 2.5 in. wire eye speculum, 4.5 in. Miltex probe, 6.5 x 6.25 x 1.25 in. storage cassette, storage cassette mat |