missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Integra™ Miltex™ Micro-Vannas Scissors

Ultra fine blades

$1,146.14 - $1,146.14

Specifications

For Use With (Application) Surgical, ophthalmic
Grade Premium
Length (English) 3.25 in.
Length (Metric) 8.3cm
Material Stainless Steel
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Blade Style Price Quantity  
Catalog Number Mfr. No. Blade Style Price Quantity  
12-460-286 Integra™ Miltex™
MLT181630
Straight Each for $1,146.14
Specifications

Description

  • High-quality surgical-grade stainless steel construction provides optimal performance
  • Fully tempered precision screw locks maintain perfect alignment
  • Ergonomic design combines weight, balance and feel for optimal comfort and performance
  • Well rounded and polished ring handles provide exceptional comfort and reduced hand fatigue
  • Length: 3.25 in. (8.3cm)

Description & Specifications

Specifications

Surgical, ophthalmic
3.25 in.
Stainless Steel
Premium
8.3cm
Micro Vannas Scissors