Please login to your online account to display your discounted pricing

Integra™ Miltex™ Mayo Scissors


Offer outstanding distal cutting capability

Manufacturer: integra™ miltex™  MLT5124TC

 View more versions of this product

Catalog No. 12-460-470

  • / Each
 

Description & Specifications

Specifications

Blade Style Straight
Material Stainless Steel
Grade Premium
Length (English) 6.75 in.
Length (Metric) 17.14cm
Type Carb-N-Sert Mayo Scissors, With standard beveled blades
For Use With (Application) Dissecting